Gene Transcripts

Gene Symbol
Ffar4
Entrez Gene ID
294075
Gene Name
free fatty acid receptor 4
Gene Aliases
Gpr120, O3far1
Chromosome Location
Chr. 1: 256786124 - 256804156 on Build Rnor_6.0
UniGene ID
Rn.82501
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047088.1 NP_001040553.1 2, 3 740
GenBank mRNA AB207868.1 - 2, 3 740

View Full Product Details