Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014146.3 | NP_054865.2 | 11 | 934 | |
NM_032463.2 | NP_115852.1 | 12 | 1053 | ||
NM_032464.2 | NP_115853.2 | 12 | 1123 | ||
XM_011516558.2 | XP_011514860.1 | 12 | 963 | ||
GenBank mRNA | AF161531.1 | - | 11 | 683 | |
AF252611.1 | - | 12 | 874 | ||
AF252612.1 | - | 10 | 629 | ||
AF252613.1 | - | 11 | 670 | ||
AF252614.1 | - | 12 | 804 | ||
AF257135.1 | - | 10 | 605 | ||
AK002099.1 | - | 12 | 913 | ||
AK092904.1 | - | 12 | 1264 | ||
AK290074.1 | - | 12 | 801 | ||
AK290916.1 | - | 11 | 682 | ||
AY190023.1 | - | 10 | 615 | ||
BC001609.1 | - | 12 | 935 | 3543310 | |
BC009204.2 | - | 10 | 607 | 4054137 |