Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300938.1 | NP_001287867.1 | 5 | 1064 | |
NM_001300939.1 | NP_001287868.1 | 5 | 1064 | ||
NM_058244.3 | NP_490645.1 | 6 | 972 | ||
NR_125351.1 | 6 | 972 | |||
XM_011543625.2 | XP_011541927.2 | 6 | 1042 | ||
XM_017009824.1 | XP_016865313.1 | 6 | 1042 | ||
XM_017009825.1 | XP_016865314.1 | 6 | 1042 | ||
XM_017009826.1 | XP_016865315.1 | 7 | 904 | ||
GenBank mRNA | AB057725.1 | - | 6 | 779 | |
AY009402.1 | - | 6 | 874 |