Gene Transcripts

Gene Symbol
Npbwr1
Entrez Gene ID
297795
Gene Name
neuropeptides B/W receptor 1
Gene Aliases
Gpr7, Nbpwr1
Chromosome Location
Chr. 5: 13379772 - 13380761 on Build Rnor_6.0
UniGene ID
Rn.140960
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014784.1 NP_001014784.1 1 229
GenBank mRNA AY577901.1 - 1 229

View Full Product Details