Gene Transcripts

Gene Symbol
Ppan
Entrez Gene ID
298699
Gene Name
peter pan homolog (Drosophila)
Gene Aliases
Chromosome Location
Chr. 8: 21905865 - 21909845 on Build Rnor_6.0
UniGene ID
Rn.144881
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011980.1 NP_001011980.1 9 974
GenBank mRNA BC089842.1 - 9 974

View Full Product Details