Gene Transcripts

Gene Symbol
CSDE1
Entrez Gene ID
7812
Gene Name
cold shock domain containing E1
Gene Aliases
D1S155E, UNR
Chromosome Location
Chr. 1: 114716913 - 114758050 on Build GRCh38
UniGene ID
Hs.69855
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007553.2 NP_001007554.1 14 2052
NM_001130523.2 NP_001123995.1 14 2106
NM_001242891.1 NP_001229820.1 15 2199
NM_001242892.1 NP_001229821.1 13 1665
NM_001242893.1 NP_001229822.1 12 1572
NM_007158.5 NP_009089.4 13 1959
GenBank mRNA AB020692.1 - 14 1966
AF077054.1 - 13 1875
AK022516.1 - 12 1474
AK022528.1 - 12 1474
AK130392.1 - 2 160
AK290146.1 - 13 1597
AL050150.1 - 3 252
AY049788.1 - 11 1461
BC032446.1 - 13 1894 5111213
BX648561.1 - 13 1796
BX648780.1 - 13 1893
CR749378.1 - 14 2056
CR749431.1 - 14 2038

View Full Product Details