Gene Transcripts

Gene Symbol
Prss32
Entrez Gene ID
302970
Gene Name
protease, serine, 32
Gene Aliases
Chromosome Location
Chr. 10: 13196700 - 13202783 on Build Rnor_6.0
UniGene ID
Rn.143939
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106983.1 NP_001100453.1 1 361

View Full Product Details