Gene Transcripts

Gene Symbol
Rab11fip4
Entrez Gene ID
303337
Gene Name
RAB11 family interacting protein 4 (class II)
Gene Aliases
RGD1305063
Chromosome Location
Chr. 10: 66942346 - 67051672 on Build Rnor_6.0
UniGene ID
Rn.224682
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107023.1 NP_001100493.1 5 772
XM_006246903.2 XP_006246965.1 5 755
XM_006246904.2 XP_006246966.1 3 717
XM_008768017.1 XP_008766239.1 5 649

View Full Product Details