Gene Transcripts

Gene Symbol
Caskin2
Entrez Gene ID
303678
Gene Name
cask-interacting protein 2
Gene Aliases
Chromosome Location
Chr. 10: 104344035 - 104358260 on Build Rnor_6.0
UniGene ID
Rn.216790
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107065.1 NP_001100535.1 3 651
XM_006247717.2 XP_006247779.1 3 689

View Full Product Details