Gene Transcripts

Gene Symbol
Rhbdf2
Entrez Gene ID
303690
Gene Name
rhomboid 5 homolog 2 (Drosophila)
Gene Aliases
Rhbdl6
Chromosome Location
Chr. 10: 105573759 - 105600885 on Build Rnor_6.0
UniGene ID
Rn.141168
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107067.2 NP_001100537.2 19 2447
NM_001270834.1 NP_001257763.1 19 2375
GenBank mRNA FQ222541.1 - 19 2375
FQ230904.1 - 19 2447

View Full Product Details