Gene Transcripts

Gene Symbol
Ttyh3
Entrez Gene ID
304315
Gene Name
tweety family member 3
Gene Aliases
Chromosome Location
Chr. 12: 16084077 - 16112664 on Build Rnor_6.0
UniGene ID
Rn.37666
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107124.1 NP_001100594.1 12 1327
XM_008768969.1 XP_008767191.1 12 1516

View Full Product Details