Gene Transcripts

Gene Symbol
Gigyf1
Entrez Gene ID
304378
Gene Name
GRB10 interacting GYF protein 1
Gene Aliases
Perq1
Chromosome Location
Chr. 12: 22234296 - 22245100 on Build Rnor_6.0
UniGene ID
Rn.206405
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107133.1 NP_001100603.1 14 1876
XM_006249150.2 XP_006249212.1 11 1638
XM_006249151.1 XP_006249213.1 11 1635
XM_006249152.1 XP_006249214.1 11 1635
XM_006249153.1 XP_006249215.1 12 1497

View Full Product Details