Gene Transcripts
- Gene Symbol
- SSPN
- Entrez Gene ID
- 8082
- Gene Name
- sarcospan
- Gene Aliases
- DAGA5, KRAG, NSPN, SPN1, SPN2
- Chromosome Location
- Chr. 12: 26122024 - 26234777 on Build GRCh38
- UniGene ID
- Hs.183428
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135823.1 |
NP_001129295.1
|
2, 3 |
284 |
|
NM_005086.4 |
NP_005077.2
|
2, 3 |
467 |
|
XM_011520853.2 |
XP_011519155.1
|
2, 3 |
217 |
|
XM_011520854.2 |
XP_011519156.1
|
2, 3 |
391 |
|
XM_011520855.2 |
XP_011519157.1
|
2, 3 |
133 |
|
GenBank mRNA |
AF016028.2 |
- |
2, 3 |
367 |
|
AK092925.1 |
- |
2, 3 |
280 |
|
AK304667.1 |
- |
3, 4 |
361 |
|
AK314353.1 |
- |
2, 3 |
456 |
|
AL136756.1 |
- |
2, 3 |
451 |
|
BC062299.1 |
- |
2, 3 |
447 |
6376868 |
BP341300.1 |
- |
2, 3 |
467 |
|
DA073350.1 |
- |
2, 3 |
284 |
|
X89105.1 |
- |
2, 3 |
289 |
|
View Full Product Details