Gene Transcripts

Gene Symbol
Snai3
Entrez Gene ID
307919
Gene Name
snail family zinc finger 3
Gene Aliases
Chromosome Location
Chr. 19: 55276302 - 55290031 on Build Rnor_6.0
UniGene ID
Rn.127187
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107439.1 NP_001100909.1 2 507
XM_006255765.2 XP_006255827.1 2 917
XM_008772611.1 XP_008770833.1 2 3930
XM_008772612.1 XP_008770834.1 3 5174

View Full Product Details