Gene Transcripts

Gene Symbol
Pcnxl2
Entrez Gene ID
307949
Gene Name
pecanex-like 2 (Drosophila)
Gene Aliases
RGD1305883
Chromosome Location
Chr. 19: 58583000 - 58736065 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006255821.2 XP_006255883.1 8 2991

View Full Product Details