Gene Transcripts

Gene Symbol
Rab11fip2
Entrez Gene ID
308003
Gene Name
RAB11 family interacting protein 2 (class I)
Gene Aliases
RGD1308538
Chromosome Location
Chr. 1: 281065346 - 281101163 on Build Rnor_6.0
UniGene ID
Rn.64225
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107447.2 NP_001100917.2 1 119
GenBank mRNA BC098050.1 - 154

View Full Product Details