Gene Transcripts

Gene Symbol
HIST1H3J
Entrez Gene ID
8356
Gene Name
histone cluster 1, H3j
Gene Aliases
H3/j, H3FJ
Chromosome Location
Chr. 6: 27890315 - 27890792 on Build GRCh38
UniGene ID
Hs.484990
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003535.2 NP_003526.1 1 301

View Full Product Details