Gene Transcripts

Gene Symbol
Lrrc4b
Entrez Gene ID
308571
Gene Name
leucine rich repeat containing 4B
Gene Aliases
Lrig4, NGL-3
Chromosome Location
Chr. 1: 100434418 - 100468914 on Build Rnor_6.0
UniGene ID
Rn.22207
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271081.1 NP_001258010.1 3 447
XM_006229055.2 XP_006229117.1 3 499

View Full Product Details