Gene Transcripts

Gene Symbol
Nipa2
Entrez Gene ID
308667
Gene Name
non imprinted in Prader-Willi/Angelman syndrome 2
Gene Aliases
RGD1306051
Chromosome Location
Chr. 1: 114346009 - 114371907 on Build Rnor_6.0
UniGene ID
Rn.2757
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107518.1 NP_001100988.1 7 1228
XM_006229292.2 XP_006229354.1 9 1331
XM_006229295.2 XP_006229357.1 7 1042
XM_006229296.2 XP_006229358.1 7 1098
XM_006229297.2 XP_006229359.1 6 1087
XM_006229298.2 XP_006229360.1 6 905
XM_006229299.2 XP_006229361.1 6 977
XM_008759390.1 XP_008757612.1 6 933
XM_008759391.1 XP_008757613.1 6 909
XM_008759392.1 XP_008757614.1 8 1492
GenBank mRNA FQ222971.1 - 6 1091

View Full Product Details