Gene Transcripts

Gene Symbol
OR1G1
Entrez Gene ID
8390
Gene Name
olfactory receptor family 1 subfamily G member 1
Gene Aliases
OR17-130, OR17-209, OR1G2
Chromosome Location
Chr. 17: 3126610 - 3127551 on Build GRCh38
UniGene ID
Hs.248183
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003555.1 NP_003546.1 1 649
GenBank mRNA BC069379.1 - 1 649
BC095520.1 - 1 680 6971960

View Full Product Details