Gene Transcripts

Gene Symbol
Mtmr10
Entrez Gene ID
309255
Gene Name
myotubularin related protein 10
Gene Aliases
RGD1305958
Chromosome Location
Chr. 1: 125124743 - 125175857 on Build Rnor_6.0
UniGene ID
Rn.3510
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100846.1 NP_001094316.1 7 743
XM_006229338.2 XP_006229400.1 7 767

View Full Product Details