Gene Transcripts

Gene Symbol
ULK1
Entrez Gene ID
8408
Gene Name
unc-51 like autophagy activating kinase 1
Gene Aliases
ATG1, ATG1A, UNC51, Unc51.1, hATG1
Chromosome Location
Chr. 12: 131894734 - 131923162 on Build GRCh38
UniGene ID
Hs.47061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003565.2 NP_003556.1 8 893
XM_011538798.2 XP_011537100.1 8 670
XM_011538799.2 XP_011537101.1 8 670
GenBank mRNA AB018265.2 - 8 675
AF045458.1 - 8 893

View Full Product Details