Gene Transcripts

Gene Symbol
Zswim6
Entrez Gene ID
310062
Gene Name
zinc finger, SWIM-type containing 6
Gene Aliases
Ab2-064
Chromosome Location
Chr. 2: 38978042 - 39042886 on Build Rnor_6.0
UniGene ID
Rn.162556
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006231909.2 XP_006231971.1 13 2461
XM_008760730.1 XP_008758952.1 10 4921
GenBank mRNA AY325185.1 - 11 1877

View Full Product Details