Gene Transcripts
- Gene Symbol
- Arhgef26
- Entrez Gene ID
- 310460
- Gene Name
- Rho guanine nucleotide exchange factor (GEF) 26
- Gene Aliases
- RGD1562559, Sgef
- Chromosome Location
- Chr. 2: 27107276 - 27236336 on Build Rnor_6.0
- UniGene ID
- Rn.7792
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_227201.8 |
XP_227201.4
|
14 |
3087 |
|
GenBank mRNA |
FQ218213.1 |
- |
13 |
1732 |
|
View Full Product Details