Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001198810.1 | NP_001185739.1 | 6, 7 | 677 | |
NM_003627.5 | NP_003618.1 | 6, 7 | 848 | ||
XM_005274358.4 | XP_005274415.1 | 7, 8 | 988 | ||
XM_017018451.1 | XP_016873940.1 | 7, 8 | 876 | ||
GenBank mRNA | AB103033.1 | - | 6, 7 | 815 | |
AF045584.1 | - | 6, 7 | 635 | ||
AK027732.1 | - | 4, 5 | 413 | ||
BC001639.2 | - | 6, 7 | 677 | 2959372 |