Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114172.1 | NP_001107644.1 | 8 | 1186 | |
NM_001303427.1 | NP_001290356.1 | 7 | 1091 | ||
NM_001303428.1 | NP_001290357.1 | 8 | 1030 | ||
NM_003629.3 | NP_003620.3 | 7 | 1685 | ||
XM_017002614.1 | XP_016858103.1 | 8 | 1063 | ||
GenBank mRNA | AK302049.1 | - | 7 | 1084 | |
AK304713.1 | - | 2 | 375 | ||
AK313726.1 | - | 8 | 1198 | ||
BC021622.1 | - | 7 | 1619 | 3910221 | |
BX537388.1 | - | 8 | 1030 | ||
CR749649.1 | - | 7 | 1606 | ||
D88532.1 | - | 7 | 1349 |