Gene Transcripts

Gene Symbol
Akap17b
Entrez Gene ID
313434
Gene Name
A kinase (PRKA) anchor protein 17B
Gene Aliases
RGD1566363
Chromosome Location
Chr. X: 123217410 - 123246049 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002730183.4 XP_002730229.1 5 2188

View Full Product Details