Gene Transcripts

Gene Symbol
Pkdcc
Entrez Gene ID
313860
Gene Name
protein kinase domain containing, cytoplasmic
Gene Aliases
RGD1311939, Sgk493
Chromosome Location
Chr. 6: 6427657 - 6436974 on Build Rnor_6.0
UniGene ID
Rn.27
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108007.1 NP_001101477.1 6, 7 1010
GenBank mRNA BC166535.1 - 6, 7 982

View Full Product Details