Gene Transcripts

Gene Symbol
Snapc1
Entrez Gene ID
314228
Gene Name
small nuclear RNA activating complex, polypeptide 1
Gene Aliases
Chromosome Location
Chr. 6: 96871675 - 96893422 on Build Rnor_6.0
UniGene ID
Rn.18361
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108033.1 NP_001101503.1 5 612
GenBank mRNA BC161969.1 - 5 631

View Full Product Details