Gene Transcripts

Gene Symbol
Plin3
Entrez Gene ID
316130
Gene Name
perilipin 3
Gene Aliases
M6prbp1, Tip47
Chromosome Location
Chr. 9: 10769739 - 10781659 on Build Rnor_6.0
UniGene ID
Rn.9506
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_236783.6 XP_236783.4 5 530
GenBank mRNA FQ230643.1 - 5 467

View Full Product Details