Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127549.1 | NP_001121021.1 | 2 | 228 | |
XM_006247724.2 | XP_006247786.1 | 2 | 830 | ||
XM_006247725.2 | XP_006247787.1 | 2 | 830 | ||
XM_006247728.2 | XP_006247790.1 | 2 | 232 | ||
XM_006247729.2 | XP_006247791.1 | 2 | 472 | ||
XM_006247730.2 | XP_006247792.1 | 2 | 232 | ||
GenBank mRNA | BC110052.1 | - | 2 | 212 | |
BC166481.1 | - | 2 | 228 | 7097977 |