Gene Transcripts

Gene Symbol
Habp4
Entrez Gene ID
361196
Gene Name
hyaluronan binding protein 4
Gene Aliases
Chromosome Location
Chr. 17: 1679562 - 1702251 on Build Rnor_6.0
UniGene ID
Rn.13726
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079940.1 NP_001073409.1 2 582
XM_006253494.2 XP_006253556.1 2 637
GenBank mRNA BC129111.1 - 2 582

View Full Product Details