Gene Transcripts

Gene Symbol
RGS9
Entrez Gene ID
8787
Gene Name
regulator of G-protein signaling 9
Gene Aliases
PERRS, RGS9L
Chromosome Location
Chr. 17: 65137338 - 65227703 on Build GRCh38
UniGene ID
Hs.664380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081955.2 NP_001075424.1 10 864
NM_001165933.1 NP_001159405.1 10 864
NM_003835.3 NP_003826.2 10 873
XM_011525426.2 XP_011523728.1 3 1044
GenBank mRNA AF071476.1 - 10 737
AF073710.1 - 1 598
AF493933.1 - 10 670
AK289876.1 - 10 737
AK290535.1 - 10 733
AK301719.1 - 10 743
AK309061.1 - 10 754
AY585191.1 - 10 661
BC022504.1 - 10 731 4801328

View Full Product Details