Gene Transcripts

Gene Symbol
Vasp
Entrez Gene ID
361517
Gene Name
vasodilator-stimulated phosphoprotein
Gene Aliases
Chromosome Location
Chr. 1: 80170608 - 80186458 on Build Rnor_6.0
UniGene ID
Rn.98750
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108475.2 NP_001101945.2 2 293
XM_008758953.1 XP_008757175.1 2 869
GenBank mRNA BC168841.1 - 2 293

View Full Product Details