Gene Transcripts

Gene Symbol
Veph1
Entrez Gene ID
361954
Gene Name
ventricular zone expressed PH domain-containing 1
Gene Aliases
RGD1308217
Chromosome Location
Chr. 2: 157869202 - 158156150 on Build Rnor_6.0
UniGene ID
Rn.158250
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014171.1 NP_001014193.1 9, 10 1956
XM_008761050.1 XP_008759272.1 8, 9 1919
XM_008761051.1 XP_008759273.1 9, 10 1956
GenBank mRNA BC087055.1 - 9, 10 1956

View Full Product Details