Gene Transcripts

Gene Symbol
Lclat1
Entrez Gene ID
362702
Gene Name
lysocardiolipin acyltransferase 1
Gene Aliases
Lycat, RGD1565906
Chromosome Location
Chr. 6: 24377338 - 24515412 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764536.1 XP_008762758.1 3 7892
XM_008764537.1 XP_008762759.1 3 442
XM_008764538.1 XP_008762760.1 3 432
XM_008764539.1 XP_008762761.1 3 7892

View Full Product Details