Gene Transcripts
- Gene Symbol
- CDC16
- Entrez Gene ID
- 8881
- Gene Name
- cell division cycle 16
- Gene Aliases
- ANAPC6, APC6, CDC16Hs, CUT9
- Chromosome Location
- Chr. 13: 114234845 - 114272675 on Build GRCh38
- UniGene ID
- Hs.374127
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001078645.1 |
NP_001072113.1
|
3, 4 |
391 |
|
NM_001318517.1 |
NP_001305446.1
|
3, 4 |
388 |
|
NM_001318518.1 |
NP_001305447.1
|
3, 4 |
388 |
|
NM_003903.3 |
NP_003894.3
|
3, 4 |
391 |
|
XM_005266206.1 |
XP_005266263.1
|
3, 4 |
433 |
|
XM_006719992.1 |
XP_006720055.1
|
3, 4 |
378 |
|
XM_006719993.1 |
XP_006720056.1
|
2, 3 |
326 |
|
XM_011534851.1 |
XP_011533153.1
|
3, 4 |
433 |
|
XM_011534853.1 |
XP_011533155.1
|
3, 4 |
433 |
|
XM_017020830.1 |
XP_016876319.1
|
3, 4 |
433 |
|
XM_017020831.1 |
XP_016876320.1
|
3, 4 |
433 |
|
XM_017020832.1 |
XP_016876321.1
|
2, 3 |
326 |
|
GenBank mRNA |
AB209850.1 |
- |
2, 3 |
1544 |
|
AF164598.1 |
- |
3, 4 |
194 |
|
AK296424.1 |
- |
3, 4 |
381 |
|
AK315302.1 |
- |
3, 4 |
222 |
|
AL540490.3 |
- |
3, 4 |
387 |
|
BC010875.2 |
- |
3, 4 |
295 |
3879150 |
BC017244.1 |
- |
3, 4 |
301 |
4877392 |
DC323610.1 |
- |
3, 4 |
394 |
|
U18291.1 |
- |
3, 4 |
214 |
|
View Full Product Details