Gene Transcripts

Gene Symbol
Pir
Entrez Gene ID
363465
Gene Name
pirin (iron-binding nuclear protein)
Gene Aliases
Chromosome Location
Chr. X: 31852322 - 31968152 on Build Rnor_6.0
UniGene ID
Rn.103685
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009474.1 NP_001009474.1 5 608
GenBank mRNA BC088290.1 - 5 608

View Full Product Details