Gene Transcripts

Gene Symbol
Tpte2
Entrez Gene ID
364629
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2
Gene Aliases
Tpte
Chromosome Location
Chr. 16: 74558529 - 74662194 on Build Rnor_6.0
UniGene ID
Rn.163336
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108877.1 NP_001102347.1 2 417

View Full Product Details