Gene Transcripts

Gene Symbol
Olr43
Entrez Gene ID
365322
Gene Name
olfactory receptor 43
Gene Aliases
Chromosome Location
Chr. 1: 167719947 - 167720897 on Build Rnor_6.0
UniGene ID
Rn.141931
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000541.1 NP_001000541.1 1 748

View Full Product Details