Gene Transcripts

Gene Symbol
TAAR5
Entrez Gene ID
9038
Gene Name
trace amine associated receptor 5
Gene Aliases
PNR
Chromosome Location
Chr. 6: 132588592 - 132589738 on Build GRCh38
UniGene ID
Hs.248198
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003967.2 NP_003958.2 1 747
GenBank mRNA AF021818.1 - 1 695
BC069171.1 - 1 695 7216924
BC095541.1 - 1 695 30706570
BC112209.1 - 1 747 8327677
BC112211.1 - 1 747 8327679

View Full Product Details