Gene Transcripts
- Gene Symbol
- Tdpoz1
- Entrez Gene ID
- 365854
- Gene Name
- TD and POZ domain containing 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 194618508 - 194619601 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_345239.7 |
XP_345240.7
|
1 |
336 |
|
View Full Product Details