Gene Transcripts

Gene Symbol
SH2D2A
Entrez Gene ID
9047
Gene Name
SH2 domain containing 2A
Gene Aliases
F2771, SCAP, TSAD, VRAP
Chromosome Location
Chr. 1: 156806238 - 156816853 on Build GRCh38
UniGene ID
Hs.103527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161441.1 NP_001154913.1 6 775
NM_001161442.1 NP_001154914.1 6 714
NM_001161443.1 NP_001154915.1 5 624
NM_001161444.1 NP_001154916.1 6 745
NM_003975.3 NP_003966.2 6 745
XM_006711615.2 XP_006711678.1 6 763
XM_017002762.1 XP_016858251.1 6 792
XM_017002763.1 XP_016858252.1 6 783
XM_017002764.1 XP_016858253.1 6 730
XM_017002765.1 XP_016858254.1 6 743
XM_017002766.1 XP_016858255.1 6 733
XM_017002767.1 XP_016858256.1 6 798
GenBank mRNA AF051325.1 - 6 667
AF097744.1 - 6 644
AJ000553.1 - 6 691
AK222737.1 - 6 668
AK313706.1 - 6 739
AY763098.1 - 5 624
AY763099.1 - 6 721
AY763100.1 - 6 651
BC012107.1 - 6 707 4636736

View Full Product Details