Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161441.1 | NP_001154913.1 | 6 | 823 | |
NM_001161442.1 | NP_001154914.1 | 6 | 762 | ||
NM_001161443.1 | NP_001154915.1 | 5 | 672 | ||
NM_001161444.1 | NP_001154916.1 | 6 | 793 | ||
NM_003975.3 | NP_003966.2 | 6 | 793 | ||
XM_006711615.2 | XP_006711678.1 | 6 | 811 | ||
XM_017002762.1 | XP_016858251.1 | 6 | 840 | ||
XM_017002763.1 | XP_016858252.1 | 6 | 831 | ||
XM_017002764.1 | XP_016858253.1 | 6 | 778 | ||
XM_017002765.1 | XP_016858254.1 | 6 | 791 | ||
XM_017002766.1 | XP_016858255.1 | 6 | 781 | ||
XM_017002767.1 | XP_016858256.1 | 6 | 846 | ||
GenBank mRNA | AF051325.1 | - | 6 | 715 | |
AF097744.1 | - | 6 | 692 | ||
AJ000553.1 | - | 6 | 739 | ||
AK222737.1 | - | 6 | 716 | ||
AK313706.1 | - | 6 | 787 | ||
AY763098.1 | - | 5 | 672 | ||
AY763099.1 | - | 6 | 769 | ||
AY763100.1 | - | 6 | 699 | ||
BC012107.1 | - | 6 | 755 | 4636736 |