Gene Transcripts

Gene Symbol
Ptges3l1
Entrez Gene ID
367808
Gene Name
prostaglandin E synthase 3-like 1
Gene Aliases
Chromosome Location
Chr. X: 45410545 - 45419817 on Build Rnor_6.0
UniGene ID
Rn.211968
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014272.1 NP_001014294.1 2 564
GenBank mRNA BC087125.1 - 2 564

View Full Product Details