Gene Transcripts

Gene Symbol
Uts2b
Entrez Gene ID
378939
Gene Name
urotensin 2B
Gene Aliases
U2B, Urp, Uts2d
Chromosome Location
Chr. 11: 76833761 - 76848808 on Build Rnor_6.0
UniGene ID
Rn.105611
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198133.2 NP_937766.1 1 127
GenBank mRNA AB116019.1 - 1 89
AY321314.1 - 1 127

View Full Product Details