Gene Transcripts

Gene Symbol
Prss58
Entrez Gene ID
408204
Gene Name
protease, serine, 58
Gene Aliases
Tryx3, try1
Chromosome Location
Chr. 4: 69145521 - 69149477 on Build Rnor_6.0
UniGene ID
Rn.125929
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009174.1 NP_001009174.1 3 344

View Full Product Details