Gene Transcripts

Gene Symbol
Mcpt4l1
Entrez Gene ID
408208
Gene Name
mast cell protease 4-like 1
Gene Aliases
Chromosome Location
Chr. 15: 34777476 - 34782348 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006252039.2 XP_006252101.2 2 211

View Full Product Details