Gene Transcripts

Gene Symbol
Vom1r77
Entrez Gene ID
494274
Gene Name
vomeronasal 1 receptor 77
Gene Aliases
V1rc37
Chromosome Location
Chr. 4: 87872330 - 87873247 on Build Rnor_6.0
UniGene ID
Rn.129663
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008926.1 NP_001008926.1 1 79
GenBank mRNA AY510297.1 - 1 79

View Full Product Details