Gene Transcripts
- Gene Symbol
- Vom1r27
- Entrez Gene ID
- 494297
- Gene Name
- vomeronasal 1 receptor 27
- Gene Aliases
- V1re19
- Chromosome Location
- Chr. 1: 68528976 - 68529893 on Build Rnor_6.0
- UniGene ID
- Rn.129439
- Species
- Rattus norvegicus
- Gene Symbol
- LOC100911644
- Entrez Gene ID
- 100911644
- Gene Name
- vomeronasal type-1 receptor 4-like
- Gene Aliases
- Chromosome Location
- Chr. 1: 68649663 - 68650566 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- Vom1r110
- Entrez Gene ID
- 494299
- Gene Name
- vomeronasal 1 receptor 110
- Gene Aliases
- V1re18
- Chromosome Location
- Chr. 1: 63184734 - 63185651 on Build Rnor_6.0
- UniGene ID
- Rn.129439
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY510356.1 |
- |
|
484 |
|
- Gene Symbol
- Vom1r29
- Entrez Gene ID
- 494292
- Gene Name
- vomeronasal 1 receptor 29
- Gene Aliases
- V1re10
- Chromosome Location
- Chr. 1: 68221395 - 68222312 on Build Rnor_6.0
- UniGene ID
- Rn.129437
- Species
- Rattus norvegicus
View Full Product Details